Gas Generating Bottle Set
Catalog #: 470123-890
Supplier: GSC International
Specifications
- Description:Gas Generating Bottle Set, Capacity: 500 mL
- Cat. no.:470123-890
- Supplier No.:9001-1
Specifications
About this item
When used with a pneumatic trough, our gas generating bottle set is ideal for generating small quantities of hydrogen and other gases.
- Hydrogen gas production requires 3M HCl and mossy zinc
The set consists of a bottle, 2-hole rubber stopper, reagent addition (thistle) tube, and gas delivery tube.
Ordering information: Pneumatic trough, ¹/₄" Int.Ø rubber tubing, and gas collection bottles are sold separately.
Customers Who Bought This Also Bought