Funnel Support
Catalog #: 470020-354
Supplier: Humboldt Manufacturing
Specifications
- Description:Funnel Support, Length 16"
- Cat. no.:470020-354
- Supplier No.:H-21650
Specifications
About this item
Featuring slotted holes for four funnels, it is constructed of hardwood with a metal clamp and thumbscrew for attaching it to a support rod.
Customers Who Bought This Also Bought