Fresnel Rectangular Magnifier
Catalog #: 470145-110
Supplier: ITOYA OF AMERICA, LTD.
Specifications
- Magnification:3×
- Description:Fresnel Rectangular Magnifier, 2 1/8" x 3 1/4"
- Cat. no.:470145-110
- Supplier No.:PL-A
Specifications
About this item
Provides Large Area of Magnification
Ultrathin fresnel lenses are flexible, unbreakable, lightweight, and washable. A protective case is included.
This credit card size magnifier can magnify up to 3X.
Customers Who Bought This Also Bought