Specifications
- Description:Ward's® Chemistry Forming Red, White and Blue Demonstration
- Cat. no.:470010-844
- Includes:Instructions
- Supplier No.:WARD470010-844
- No. of Uses:7
Specifications
About this item
A Great Way to Introduce Chemical Reaction Types
- Provide for interesting discussion of 3 different chemical reactions
This “patriotic” demonstration illustrates complex ion formation, double replacement reaction and Prussian blue. Three reactions are carried out in adjacent beakers resulting in red, white and blue solutions.