Foraminifera Thin Section
Catalog #: 470026-822
Supplier: Avantor
Specifications
- Description:Foraminifera thin section
- Supplier No.:WARD470026-822
Specifications
About this item
Cross section of a Pennsylvanian fusulinid in chert that shows random orientations of structural features, including the proloculus and chamber walls.
Customers Who Bought This Also Bought