Fluorescent Viewing Goggles
Catalog #: 470019-920
Supplier: NOIR MEDICAL
Specifications
- Description:Fluorescent Viewing Goggles
- Cat. no.:470019-920
- Supplier No.:U60
Specifications
About this item
Necessary Protection When Operating Forensic Light Source
Completely shield your eyes from UVA and UVB rays with these flexible orange viewing goggles. Clearly see
latent prints and other evidence fluorescing with our wraparound goggles that meet ANSI standards and comfortably fit over prescription glasses.
Customers Who Bought This Also Bought