Ward's® Exploring Stellar Light and Temperature Lab Activity
Catalog #: 470023-714
Supplier: Avantor
Specifications
- Description:WARD’S Exploring Stellar Light and Temperature Lab Activity, Materials for Ten Lab Groups
- Cat. no.:470023-714
- Supplier No.:WARD470023-714
Specifications
About this item
Analyze Star’s Spectra and Calculate Temperatures
Students learn to use a diffraction grating with a variable resistance light board to observe how light is separated into various colors, then determine how a star’s color indicates its temperature. It comes with a teacher’s guide and student copymasters.
Customers Who Bought This Also Bought