Ward's® Live Euglena sp. (gracilis)
Supplier: Ward's Science
Certificates
About this item
Characterized by a lack of rigid cellulosic walls, instead these organisms have pellicles made of protein. They store paramylon instead of starch.
- Excellent for studying general flagellates
Study Set contains: a culture of Euglena sp. And a Form and Structure Copymaster diagram
Ordering information: These products contain living or perishable materials and will ship via overnight or 2nd day delivery.
Delivery: Please specify a delivery date during checkout.