Essential Physics Demo: Venturi Tube
Catalog #: 470018-310
Supplier: VWR International
Specifications
- Description:Essential Physics Demo: Venturi Tube
- Size:45 x 13 x 32 cm
- Cat. no.:470018-310
- Supplier No.:LIQ_DEM
Specifications
About this item
Investigate the relationship between the speed of a fluid and its pressure
- Easy to use
This impressive demonstration consists of a glass Ventri tube with two different diameters, with a set of additional tubes attached to the different sections . One pair holds a U-shaped tube in which a color liquid can be placed to demonstrate the different pressures exerted when air is blown through the tube.
Ordering information: Demo includes two rubber hoses, glass Venturi tube, U-shaped tube and two hose clamps.
Customers Who Bought This Also Bought