Ectocarpus
Catalog #: 470177-290
Supplier: CONNECTICUT VALLEY BIO SUPPLY
Specifications
- Description:Ectocarpus (Small)
- No. of Students:30
- Cat. no.:470177-290
- Packaging:Tube
- Media:Erdschreiber's medium
- Supplier No.:WARD470177-290
Specifications
About this item
Marine. Filamentous. Branched thallus, zoospores. Erdschreiber’s medium.
Kelp and seaweed; with a distinct set of photosynthetic pigments.
Customers Who Bought This Also Bought