G.S.C. Spark Timers
Supplier: G.S.C. INTERNATIONAL, INC.
Certificates
About this item
The Go Science Crazy spark timer is a recording device used to measure the velocity and acceleration of a falling or moving object.
- Spark timer device
- 60 Hz (mains power frequency)
- Tape length: 45 m
- Tape width: 17 mm
The Go Science Crazy spark timer sparks at uniform intervals, leaving marks on a paper tape. The distance between the marks on the tape are used to calculate velocity and acceleration. It is provided with a roll of tape, a carbon disc, a weight, and a clamp to hold it in position.
Everything on the spark timer is enclosed for safety.
Delivery: Timer includes five carbon disks, one 45 m roll of tape, clamp, and weight. Additional rolls of tape and carbon disks are also available separately.
Customers Who Bought This Also Bought