Economy Optics Kit
Catalog #: 470144-284
Supplier: Shiv Dial Sud & Sons
Specifications
- Description:Economy Optics Kit
- Cat. no.:470144-284
- Supplier No.:OPTKIT-B
Specifications
About this item
Everything You Need to Demonstrate Optical Principles
This kit contains one double convex lens, one double concave lens, one plain lens, one glass screen, two mirrors, three supports, five cellophane squares of different colors, two equilateral prisms, one beaker, one flashlight, one plastic screen, and one candle.
Customers Who Bought This Also Bought