Double-Walled Calorimeter
Catalog #: 470018-276
Supplier: PRECISION MOULDED POLYMERS LIMITED
Specifications
- Description:Double-Walled Calorimeter
- Cat. no.:470018-276
- Supplier No.:M40101
Specifications
About this item
Simple Calorimeter to Measure Heat in Many Applications
The double-walled cover minimizes heat loss, and the inner and outer vessels are both made of aluminum. The inner vessel, which has a 200 mL capacity, comes with a stirrer and support.
Customers Who Bought This Also Bought