Specifications
- Description:DNA Fingerprinting I Lab Activity
- No. of Lab Groups:8
- Cat. no.:470200-476
- Supplier No.:109
Specifications
About this item
In this experiment, crime scene DNA fingerprints are compared with those obtained from suspects.
- Required Components Included
- Time Required: 45 minutes
- Kit Explains Basic Concepts of DNA Fingerprinting
DNA fingerprinting is a method used in various forensic procedures as well as paternity determinations. The experiment provides the basic reagents and biologicals for DNA fragmentation pattern analysis and does not contain human DNA.
Horizontal gel electrophoresis apparatus, DC power supply, and micropipettes with tips are required but not included.