Dinobot Hydraulic Robot Kit
Catalog #: 470106-076
Supplier: Avantor
Specifications
- Description:Kit, Dinobot Hydraulic Robot
- Cat. no.:470106-076
- Supplier No.:WARD470106-076
Specifications
About this item
Students build their own working hydraulic robot.
They can either use the included dinosaur pattern or come up with a creation of their own design. A great way to explore concepts of robots and introduce hydraulics. Robot has three sets of controls.
Kit includes 60mL syringes, plastic tubing, food coloring, base and syringe holders, brass fasteners, foamcore, and instructions. Developed by George Wright.
Customers Who Bought This Also Bought