Ward's® Dichotomous Key To Free-Living Protists
Catalog #: 470190-778
Supplier: Ward's Science
Specifications
- Description:Protists Chart
- Cat. no.:470190-778
- Supplier No.:DEFAULT VPN
Specifications
About this item
Identify Over Thirty Different Forms
Guiding students through descriptions of protists and providing illustrations of a number of organisms, our dichotomous key helps students identify unknown types they observe in the lab.
Customers Who Bought This Also Bought