Cube-O-Gram Cubes
Catalog #: 470014-880
Supplier: PRECISION MOULDED POLYMERS LTD
Specifications
- Description:Cube-O-Gram Cubes
- Cat. no.:470014-880
- Supplier No.:A-M 1050
Specifications
About this item
Introduce a variety of basic scientific concepts and properties.
It's easy and fun for students to learn the metric system, conservation of mass, factoring, the Pythagorean Theorem, and much more with the colorful, building-block-style Cube-O-Grams. These colorful, 1 cubic centimeter pieces each have a volume of 1 mL and mass of 1 g; snap them together in any number of ways to illustrate virtually any scientific concept you can think of. Package of 100 cubes.
Caution: WARNING: CHOKING HAZARD- Small Parts. Not for Children Under 3 Years.
Customers Who Bought This Also Bought