Ward's® Crystal Systems Reference Slide Set
Catalog #: 470183-428
Supplier: Avantor
Specifications
- Description:WARD’S Crystal systems reference slide set, set of 6
- Cat. no.:470183-428
- Supplier No.:WARD470183-428
Specifications
About this item
Explore the characteristics of each of the six crystal systems.
An ideal supplement to your class's work with crystal models, WARD'S permanent reference set contains standard 1×3" glass microscope slides of real crystals for examining the different crystal forms. You will also receive a study guide. Set of six.
Set includes the following crystals: monoclinic, orthorhombic, hexagonal, tetragonal, isometric, and triclinic.
Customers Who Bought This Also Bought