Crucible Cradle
Catalog #: 470202-756
Supplier: Global Market Connections
Specifications
- Length:101/2"
- Description:Crucible Cradle
- Cat. no.:470202-756
- Supplier No.:CC-2006
Specifications
About this item
Introducing the safest way to heat, cool, and weigh a crucible; this innovative device allows students to handle porcelain crucibles at extremely high temperatures without any direct contact.
- Easy to maneuver
- Long, strong handle
- The cradle can be placed on or in a heating surface and on a scale
- Eliminates the need for awkward crucible tongs
- Ultimately makes your laboratory experiments safer
A sturdy wooden handle is attached to a metal ring with porcelain pipe stem triangular supports, which steadily secure the crucible.
Customers Who Bought This Also Bought