Crayola Crayons (Pkg/24)
Catalog #: 470104-042
Supplier: EATON OFFICE SUPPLY
Specifications
- Color:24 assorted
- Description:Crayola Tuck Box
- Cat. no.:470104-042
- Supplier No.:BIN50-3024
Specifications
About this item
The classic box of crayons for every use. T
he box is small enough to fit in a book bag or knapsack, but large enough to hold a wide variety of colors. From carnation pink to cerulean, indigo to scarlet, you can mix and match to create colorful drawings and bold signs. It's a must-have for every kid!
Contains 24 non-toxic crayons. Bright and vivid colors.
Customers Who Bought This Also Bought