Connecting Wires (BNC to Alligator Clips)
Catalog #: 470122-140
Supplier: Avantor
Specifications
- Description:Connecting wires (BNC to alligator clips)
- Cat. no.:470122-140
- Cord Length:18"
- Supplier No.:4531-C-18
Specifications
About this item
Flexible, rubber-covered wire with covered plated steel alligator clips on ends.
Wire is stranded. Set of two leads: one black, one red.
Customers Who Bought This Also Bought