Comprehensive Mineral and Physical Properties Collection
Catalog #: 470112-328
Supplier: SCOTT RESOURCES 2, LLC SE
Specifications
- Description:Comprehensive Mineral Collection
- No. of Samples:126
- Cat. no.:470112-328
- Supplier No.:2340
Specifications
About this item
Teach students how to identify minerals with this collection.
- Covers multiple mineral properties
- Specimen lists included
Teach your students how to identify minerals by their physical properties with this extensive 126-sample collection. Each 1-1/2" x 1-1/2" specimen was chosen to show the following characteristics: cleavage and fracture, luster, hardness and tenacity, color, streak crystalline aggregates, fluorescence (short and long wave), specific gravity, other properties such as optical, thermal and magnetic. Specimens are packed in compartmented trays with a list giving identity and sample origin.
Customers Who Bought This Also Bought