Compound Hand Magnifier
Catalog #: 470105-656
Supplier: United Scientific Supplies
Specifications
- Magnification:2×, 6×, 8×
- Description:Compound Hand Magnifier
- Cat. no.:470105-656
- Supplier No.:PTM-003
Specifications
About this item
Two or more magnifiers may be snapped together to produce increased magnification and to illustrate the principles of compound lenses.
- Three magnifiers in one
- All plastic
Size: 100 mm.
Delivery: Each magnifier includes 2x, 6x, and 8x lenses constructed of optical grade plastic.
Customers Who Bought This Also Bought