About this item
This organism can be isolated from soil, wounds, and intestinal tracts of humans. It digests protein and is known for its strong, distinctive odor.
- Biosafety Level: 1
- Gram Stain: Positive
- Morphology: Rods
- Motility: Positive
- Respiration: Anaerobic
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in a freeze-dried culture for longer storage.
Freeze-dried organisms come with everything you need to rehydrate and culture the organism when you're ready.
Bacterial and fungal cultures are restricted and can only be ordered by educational and research professionals.
Ward's Science is not affiliated with the American Type Culture Collection and does not provide corresponding ATCC numbers for our organisms.