Ward's® Cleavage Collection
Catalog #: 470025-248
Supplier: Avantor
Specifications
- Description:Cleavage Collection
- No. of Samples:6
- Cat. no.:470025-248
- Supplier No.:WARD470025-248
Specifications
About this item
Specimens Illustrate Smooth Mineral Breakage
- Hand picked by WARD's Geologists
- Specimen list included
- Teaches students how minerals natural cleavage
Each sample shows a different type of cleavage that leaves a smooth surface. The set is housed in a compartmented collection box, and an identification list is included.
Ordering information: Contents include 6 numbered samples (1" x 1"), Basal Cleavage (Mica), Pinacoidal Cleavage (Feldspar), Prismatic Cleavage (Amphibole), Cubic Cleavage (Halite), Rhombohedral Cleavage (Calcite), and Octahedral Cleavage (Fluorite).
Customers Who Bought This Also Bought