Classroom PCR LabStation
Catalog #: 470122-884
Supplier: Edvotek
Specifications
- Description:Classroom PCR LabStation for up to 25 students
- Cat. no.:470122-884
- Supplier No.:5067
Specifications
About this item
A complete package for your classroom PCR needs.
- Supports up to 25 students
- Includes all materials necessary to perform PCR assays start to finish
Kit includes: 1× EdvoCycler™ 2 (48×0.2 ml); 6× M12 Complete™ package (14×7 cm tray and 7×7 cm trays (2)); 3 x model 509 DuoSource™ Power Supplies (100 or 150 V, for 1 to 2 units); 6× Variable MicroPipette (5 to 50 µl); 1× TruBlu™ 2 LED transilluminator and 1× 3 L water bath.
Customers Who Bought This Also Bought