Ward's® Live Chlamydomonas moewusii Cultures
Supplier: Ward's Science
Certificates
About this item
Demonstrates isogamous reproduction where gametes are similar in appearance and behavior. Make lectures and labs on protist reproduction clearer with our specially selected cultures that illustrate various reproductive phenomona.
- Both mating types (+) and (-) available
- Bacteria-free (Axenic) cultures available for both mating types
This green alga is a unicellular flagellate commonly found in damp soil, and aquatic environments . Each cell has 2 flagella and a large eyespot that allow it to sense light.
Chlamydomonas moewusii can only survive in high levels of light, while C. reinhartdii (also available) can also survive in the dark if it is provided organic carbon.
Genus: Chlamydomonas Species: moewusii
Related Information
Algae_and_Cyanobacteria.pdf Working_with_Algae_and_Cyanobacteria.pdf
Product Literature
Customers Who Bought This Also Bought