TeacherGeek Check Valve - ¹/₈"
Catalog #: 470132-476
Supplier: TEACHERGEEK INC
Specifications
- Description:Check Valve - 1/8", Pkg. of 10
- Cat. no.:470132-476
- Supplier No.:44101
Specifications
About this item
Check valves allow one way passage of water or air through tubing.
Check valves allow one way passage of water or air through tubing. They are used in pumps, jacks and other hydraulic/pneumatic applications.
They are used in pumps, jacks and other hydraulic/pneumatic applications.
Customers Who Bought This Also Bought