Cervix Uteri, Section, Hematoxylin and Eosin Stained Slides
Catalog #: 470183-114
Supplier: Avantor
Specifications
- Description:Cervix Uteri
- Cat. no.:470183-114
- Stain:Hematoxylin and Eosin
- Supplier No.:WARD470183-114
- Orientation:Section
Specifications
About this item
Demonstrates Varying Tissue Types
- Identifying features clearly distinguishable
- Rigorous quality control standards
Human. Columnar epithelium in the canal of the cervix; stratified squamous epithelium on external part of portio vaginalis cervicus.
Customers Who Bought This Also Bought