Centripetal Force Kit
Catalog #: 470149-398
Supplier: OSAW INDUSTRIAL PRODUCTS SE
Specifications
- Description:Centripetal Force Kit
- Cat. no.:470149-398
- Supplier No.:SV583
Specifications
About this item
The Centripetal Force Kit is inexpensive and effective.
- Designed for PSSC
- Plastic Tube
- Two Rubber Stoppers
- Steel Washers
- Instructions Included
This kit was originally designed for PSSC, and it provides excellent data with centripetal force.
A plastic tube works as the pivot, two rubber stoppers are the whirling mass, steel washers are the counterforce, and the nylon cord ties the stoppers to the washers.
Ordering information: This kit includes complete instructions.
Customers Who Bought This Also Bought