Specifications
- Description:Cells of plants microslides
- Cat. no.:470008-986
- Supplier No.:T-11
Specifications
About this item
Investigate plant-specific cells.
- Arrows help students locate important features
- Mounted in a clear plastic holder that protects it on both sides
- Set of 15 includes detailed lesson plan on the topic under study
Because plants do not move about the way animals do, students sometimes forget that they are living creatures too. This lesson examines plants at the cellular level, showing students a few of the different kinds of cells there are in plants and the special functions they perform.
The set features one strip of eight images. Also included is a key to image descriptions.
Introduce junior and senior high school students to the world of microscopy with inexpensive, unbreakable Microslides. Microslides consist of a strip of film with eight photomicrographs, each reproduced exactly as you would see them through a professional microscope under ideal conditions. You can supply all your students with their own Microslides. A detailed text folder is included. Viewers are available separately.
Contents:
Onion skin cells - wm/st (110×)
Green cells of a leaf - wm (150×)
Cells of a woody stem - ls/st (100×)
Cells of a woody stem - cs/st (40×)
Cells of the anther - cs/st (100×)
Cells of the lily ovary - ls/st (300×)
Spirogyra cell - wm (175×)
Bacteria - wm/st (800×)