Cells
Catalog #: 470111-848
Supplier: Kemtec
Specifications
- Description:Cells I Kit
- No. of Lab Groups:6
- Cat. no.:470111-848
- Supplier No.:1-152
Specifications
About this item
Learn about cells like Robert Hooke.
- Make your own cell slides
- Study plant and animal cells
The emphasis is on cell structure theory as students view thin slices of cork cells in a quest for knowledge on cell structure and cell organelles. Plant cells include onion and potato cells. Animal cells include cheek cells and microorganisms from pond water.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought