Celestron PowerSeeker 60 Refractor Telescope
Supplier: Celestron International
Certificates
About this item
Our most economical telescope.
- Simple, affordable, and effective
- Supported by "The Sky" astronomy software
- Includes a preassembled tripod and accessory tray
An ideal combination of quality, value, features, and power, this telescope is well-equipped for both celestial and terrestial viewing. Optical performance is provided from a 3X Barlow lens, tripling the magnifying power of each eyepiece. Easy to assemble, the unit comes with pre-assembled aluminum tripod and accesssory tray. Telescope includes a 5 x 24 finderscope, and “The Sky” astronomy software.
Features:
• 3X Barlow lens
• 700 mm focal length; ƒ11.67
• 4 mm and 20 mm eyepieces
• 35X or 175X magnification
• Weight: 7 lbs.
Customers Who Bought This Also Bought