Ward’s® Live Black Planaria (Phagocata gracilis)
Supplier: Avantor
Certificates
About this item
Black Planaria are a flatworm commonly used in science fair projects. They do not bite and move very slowly, making them easy for young students to observe.
- Study structure, feeding habits, responses to environmental stimuli and regeneration
- Compare to Brown and White Planaria (also available)
This species is similar to Brown Planaria but can grow slightly larger and has a ruffled plicate pharynx.
Black planaria are up to 1/2" in size.
Ordering information: These products contain living or perishable materials.
Delivery: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Customers Who Bought This Also Bought