Specifications
- Description:Bio-Plastic™ Deluxe Gallon Package
- Cat. no.:470191-728
- Supplier No.:BB0117-1GKIT
Specifications
About this item
Think of how many mounts could be made with this package! It comes with a total of one gallon of Bio-Plastic in eight 1 pint bottles along with eight vials of catalyst.
Bio-Plastic should be stored in a cool, dry area and has a shelf life of six months.
We are required to ship all Bio-Plastic and accompanying Bio-Plastic chemical compounds via UPS to insure safe, timely delivery.