Specifications
- Description:Bio-Energy Education Kit
- No. of Lab Groups:1
- Cat. no.:470145-654
- Supplier No.:FCJJ-22-B
Specifications
About this item
Have your students explore the alternatives to hydrogen and methanol as fuel with this kit.
Have your students explore the alternatives to hydrogen and methanol as fuel with this kit. Using ethanol as a fuel source, this kit silently converts ethanol (alcohol) to electricity without any combustion. The device provides for an exemplary demonstration of next-generation bio-fuel cell technology as it runs non-stop for hours. This kit includes a renewable energy education manual and an experiment guide.
Restriction Comments:
This kit is intended only for use by person 12 years old and up, and only under the supervision of an adult who has read and understood the instructions provided in the kit’s user manual.
Warning: Choking Hazard: Not suitable for children under 3 yrs.
- 12 years old and up
- Ethanol-powered fuel cell
- Includes an education manual and experiment guide
Using ethanol as a fuel source, this kit silently converts ethanol (alcohol) to electricity without any combustion. The device provides for an exemplary demonstration of next-generation bio-fuel cell technology as it runs non-stop for hours. This kit includes a renewable energy education manual and an experiment guide.
Restriction Comments:
This kit is intended only for use by person 12 years old and up, and only under the supervision of an adult who has read and understood the instructions provided in the kit’s user manual.
Warning: Choking Hazard: Not suitable for children under 3 yrs.