Bile Esculin Agar
Catalog #: 470180-632
Supplier: IPM SCIENTIFIC, INC.
CAS Number:
Specifications
- Description:Dehydrated Bile Esculin Agar
- Cat. no.:470180-632
- Supplier No.:880010
Specifications
About this item
For differential isolation and presumptive identification of group D streptococcus.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.
Customers Who Bought This Also Bought