Basic Power Supply Accessory Kit
Catalog #: 470201-004
Supplier: CAL TEST ELECTRONICS SE
Specifications
- Description:Basic Power Supply Accessory Kit
- Cat. no.:470201-004
- Supplier No.:CT3374
Specifications
About this item
Useful tools for any power supply.
This basic kit provides the necessary test leads and alligator clips required when starting to use your general-purpose bench-top AC or DC power supply. The kit features leads with high capacity silicone jacketed wire with retractile sleeve to straight fixed sheathed banana plugs. Two insulated alligator clips make the final connections. The four piece kit comes package in a clear PVC pouch for convenient storage. RoHS compliant.
Customers Who Bought This Also Bought