Fundamentals of Physics Atomic Spectra Bench
Catalog #: 470017-102
Supplier: VWR International
Specifications
- Description:Atomic Spectra Bench
- No. of Lab Groups:1
- Cat. no.:470017-102
- Supplier No.:FP-ASPRE
Specifications
About this item
This fun and interactive kit enables students to research atomic spectra and spectral line analysis.
- Teacher's Manual Included
- Students Calculate Colored Rods' Wavelength
- Colored Rods Included
- Interactive Activities
- Purpose for Students: to Understand Atomic Spectra and Spectral Line Analysis
With the FUN stand on its side, teachers can place colored rods at different locations, so students can observe the atomic spectra from a gas tube.
Students can also calculate the wavelength of each colored rod based on its distance from the central axis with metric tape and diffraction grating.
Ordering information: This kit includes colored rods, a support stand, instructions, metric tape, and diffraction grating. A gas tube, power supply, and Fun stand are required but not included.
Customers Who Bought This Also Bought