Aspirator Filter Pump
Catalog #: 470202-842
Supplier: Humboldt Manufacturing
Specifications
- Description:Aspirator Filter Pump
- Cat. no.:470202-842
- Supplier No.:H-12020
Specifications
About this item
Ideal for vacuum filtrations under both high and low water pressure.
- A ball-type, positive action valve in the suction tube prevents backflow if pressure is reduced
- Nickel-plated and designed for greater air entrainment capacity
- Chapman, Swirling-type
- 3/8" NPT tapered
- Size: 5.375" L
Customers Who Bought This Also Bought