Amino Acids SuperModel Kit
Catalog #: 470029-876
Supplier: RYLER ENTERPRISES, INC.
Specifications
- Description:Amino Acids SuperModel Kit
- Cat. no.:470029-876
- Supplier No.:AA-2
Specifications
About this item
Teach protein structure with models and activities.
- Construct all 20 amino acids in both the L and D forms
- Includes connectors for single and double bonds
- Instruction booklet
You can construct all 20 amino acids in both the L and D forms, as well as make peptide bonds and polypeptides with these flexible and durable model parts. The kit includes 29 carbons, 16 oxygens, 54 hydrogens, three sulfurs, 11 nitrogens, and 109 connectors for single and double bonds, as well as an instruction booklet.
Customers Who Bought This Also Bought