LaMotte® Alkalinity Water Test Kit
Supplier: LaMotte
Certificates
About this item
Precalibrated test tubes and labware give fast, accurate results on-site, in the classroom, or in the lab.
- Range: 0 to 200 ppm
- Materials for 50 tests
- Aligned with GLOBE protocols
Reagents, labware, and step-by-step instructions are furnished in a compact, waterproof carrying case.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.