Specifications
- Description:Air Track, Complete w/3 Gliders (2 Boxes)
- Cat. no.:470007-436
- Supplier No.:SH501A
Specifications
About this item
With the Air Track, students can investigate motion phenomena at a precise level of measurement.
Constructed to the demanding standards typical of more expensive units, this air track offers students the possibility of investigating motion phenomena at a precise level of measurement. The sturdy extruded black aluminum construction of the track and the two included gliders have been designed with easy setup in mind. Quick-release set screws make fitting the many included accessories very easy. The comprehensive accessory set includes two sets of photogate flags, two elastic collision springs, an inelastic collision device, three vehicles: two larger weighing 400g and one smaller weighing 200g, magnets and velcro pads, counterweight. An air source (not included) is required for operating the air track. Size: 200 x 20 x 14cm.
- Easy Setup
- Quick-Release Set Screws
- Two Sets of Photogate Flags
- Two Elastic Collision Springs
- One Inelastic Collision Device
The sturdy, extruded black aluminum construction of the track and the two included gliders have been designed with easy setup in mind. Quick-release set screws make setting up the multiple provided accessories easy.
The Air Track measures 200 x 20 x 14 cm.