Adjustable Micro Bunsen Burners
Catalog #: 470148-840
Supplier: VWR International
Specifications
- Description:Adjustable micro Bunsen burners
- Cat. no.:470148-840
- For:Natural gas
- BTU Output:800 - 1200 BTU/cu. ft.
- Supplier No.:C-1486
Specifications
About this item
Ideal for Jobs Requiring a Small, Intense Flame
- Nickel-plate, die-cast zinc alloy base
- Flame stabilizer and needle valve for precise flame control
- Gas inlet is serrated for secure connections
Gas inlet 5/16" outer dia.; mixing tube 7/16" outer dia.; 3 7/8"H
Customers Who Bought This Also Bought