Ward's® Manipulate Your Own Action Potential Neuron
Catalog #: 470105-828
Supplier: Avantor
Specifications
- Description:Action Potential Neuron kit
- No. of Lab Groups:15
- Cat. no.:470105-828
- Supplier No.:WARD470105-828
Specifications
About this item
Students use the manipulatives in this activity kit, developed by Teacher Carole MacMullan, to depolarize and repolarize a neuron as they model sodium and potassium ion movement through gated channel proteins embedded in the plasma membrane of a neuron.
- Hands-on learning helps solidify an abstract concept
- Suitable for grades 10-12
Students manipulate, then restore the neuron to its resting potential state with the help of ATP and a sodium/potassium pump.
Customers Who Bought This Also Bought