Ward's® Chemistry Acid-Fast Stain Kit
Catalog #: 470038-940
Supplier: Ward's Science
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Ward's® Acid-Fast Stain Kit, For 50 Slides
- Cat. no.:470038-940
- Supplier No.:9516700
Specifications
About this item
Identify the family Mycobacteriaceae by their ability to retain a fuschia color following the staining procedure.
Delivery: Includes carbol-fuschin solution, methylene blue solution, acid alcohol destain, and instructions.
Customers Who Bought This Also Bought