SP Bel-Art Reagent/Acid Pump Dispenser, Bel-Art Products, a part of SP
Catalog #: 470103-610
Supplier: Bel-Art
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Scienceware® reagent/acid pump dispenser
- Cat. no.:470103-610
- Supplier No.:F116560000
Specifications
About this item
Aids in the transfer of acids and reagents and virtually eliminates accidental bottle breakage and spills.
- Squeeze the small plastic bottle to begin flow; press the pressure release button to immediately stop dispensing
- Fits many 5 pint (80 fl. oz.) acid bottles, ½ and 1 gallon jugs, 2.5 and 4 liter bottles, and similar bottles with GPS No. 38-430 or 38-400 threaded necks (not included)
- Two adapters provided for short necks
Customers Who Bought This Also Bought