7-Place Wood Test Tube Support
Catalog #: 470149-434
Supplier: VWR International
Specifications
- Description:7-Place Wood Test Tube Support
- Cat. no.:470149-434
- Supplier No.:CT1618
Specifications
About this item
With drilled holes to accommodate seven tubes up to 13 mm in dia. Strongly constructed with dovetail joints. Size: 2-3/8" H x 7-7/8 W x 1-3/8" D.
Customers Who Bought This Also Bought