ContactUsLinkComponent
60-Minute Timer
60-Minute Timer
Catalog #: 470017-312
TIMER 60 MINUTE
60-Minute Timer
Catalog #: 470017-312
Specifications
  • Description:
    60-minute timer
  • Cat. no.:
    470017-312
  • Supplier No.:
    TI030001
Specifications
About this item
This economical, compact timer measures up to one hour in one-minute intervals.
It is spring-powered and requires no batteries.
Customers Who Bought This Also Bought