Specifications
- Description:Fireproof vertical filing cabinet
- Cat. no.:470116-200
- Type:2 drawers
- Supplier No.:4-1825-C
Specifications
About this item
"Fireproof insulation is 100% gypsum, reinforced by a 1" by 2" lattice made of 14-gauge galvanized welded steel wire. Field replaceable steel panels can be easily replaced. Finish is an environmentally friendly, scratch-resistant electrostatic powder coating applied to all sides, including bottom. Drawer bodies are built with high sides for use with hanging folders.
- 2-position drawer catch allows access to certain drawers while others remain locked
Drawer locks on all files provide general locking of all drawers through either standard key locks or UL-listed high-security locks. Drawer pulls are surface mounted to allow for extra insulation inside the drawer heads. Insulation between all drawers makes each drawer a separate insulated container.Drawer heads are formed of welded steel and filled with fireproof gypsum insulation and are field replaceable.